The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007200.1 | NP_001007201.1 | 17 - 18 | 1662 | 103 |
GenBank mRNA | BC162720.1 | - | 17 - 18 | 1676 | 103 |