Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt2
Entrez Gene ID
100005717
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
wu:fa03e08, wu:fb54a02, zgc:101627
Chromosome Location
Chr.8: 174143 - 179265 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007447.1 NP_001007448.1 3 - 4 500 88
GenBank mRNA BC085392.1 - 3 - 4 500 88
BC164562.1 - 2 - 3 295 88

View Full Product Details