Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nxnl2
Entrez Gene ID
494087
Gene Name
nucleoredoxin like 2
Gene Aliases
zgc:101722
Chromosome Location
Chr.10: 2582254 - 2585625 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC086727.1 - 2 - 3 789 93

View Full Product Details