Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fut10
Entrez Gene ID
497615
Gene Name
fucosyltransferase 10
Gene Aliases
zgc:195147
Chromosome Location
Chr.10: 7255533 - 7264913 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012374.1 NP_001012374.1 1 - 2 99 73
GenBank mRNA AJ879586.1 - 1 - 2 99 73

View Full Product Details