Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc29a2
Entrez Gene ID
503537
Gene Name
solute carrier family 29 member 2
Gene Aliases
zgc:110527
Chromosome Location
Chr.21: 27407490 - 27413270 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012501.2 NP_001012519.2 5 - 6 479 117
GenBank mRNA BC076404.1 - 5 - 6 480 117
BC090184.1 - 5 - 6 479 117

View Full Product Details