Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mybpc2b
Entrez Gene ID
541384
Gene Name
myosin binding protein Cb
Gene Aliases
im:7137115, im:7150089, zgc:110761
Chromosome Location
Chr.24: 38259520 - 38348808 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013511.2 NP_001013529.2 7 - 8 482 69
XM_017353828.2 XP_017209317.1 6 - 7 547 69
XM_021470159.1 XP_021325834.1 7 - 8 516 69
XM_021470160.1 XP_021325835.1 6 - 7 544 69
XM_021470161.1 XP_021325836.1 6 - 7 494 69
XM_021470162.1 XP_021325837.1 6 - 7 431 69
GenBank mRNA BC091662.1 - - 476 69
CK676323.1 - 7 - 8 476 69
EB790963.1 - 3 - 4 249 69

View Full Product Details