Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gcna
Entrez Gene ID
541448
Gene Name
germ cell nuclear acidic peptidase
Gene Aliases
acrc, zgc:113346
Chromosome Location
Chr.14: 14667274 - 14687035 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013573.1 NP_001013591.1 7 - 8 666 140
XM_021481013.1 XP_021336688.1 7 - 8 693 140
XM_021481015.1 XP_021336690.1 7 - 8 693 140
XM_021481016.1 XP_021336691.1 7 - 8 715 140
GenBank mRNA BC090463.1 - 7 - 8 666 140
BC154362.1 - 7 - 8 687 140

View Full Product Details