Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trim2a
Entrez Gene ID
100003782
Gene Name
tripartite motif containing 2a
Gene Aliases
trim2, zgc:113099
Chromosome Location
Chr.1: 25322445 - 25370281 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014371.1 NP_001014393.1 8 - 9 1919 110
XM_005170889.4 XP_005170946.1 8 - 9 1998 110
XM_005170890.4 XP_005170947.1 4 - 5 1091 110
GenBank mRNA BC091789.1 - 8 - 9 1919 110

View Full Product Details