Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crlf3
Entrez Gene ID
550515
Gene Name
cytokine receptor-like factor 3
Gene Aliases
fd20c08, id:ibd2508, im:7137736, wu:fd20c08, zgc:110212
Chromosome Location
Chr.3: 35812049 - 35847317 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017817.2 NP_001017817.2 3 - 4 421 116
GenBank mRNA AM233512.1 - 3 - 4 418 116
BC092800.1 - 3 - 4 416 116
BC155810.1 - 3 - 4 401 116

View Full Product Details