Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
baiap2l1b
Entrez Gene ID
550528
Gene Name
BAR/IMD domain containing adaptor protein 2 like 1b
Gene Aliases
zgc:110192
Chromosome Location
Chr.3: 61401311 - 61494850 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017830.1 NP_001017830.1 12 - 13 1572 57
XM_009299879.3 XP_009298154.1 12 - 13 1588 57
GenBank mRNA BC092786.1 - 12 - 13 1572 57

View Full Product Details