Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs16
Entrez Gene ID
569828
Gene Name
regulator of G protein signaling 16
Gene Aliases
zgc:110164
Chromosome Location
Chr.6: 189855 - 192556 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017846.1 NP_001017846.1 1 - 2 106 67
GenBank mRNA BC092767.1 - 1 - 2 106 67
BC164299.1 - 1 - 2 101 67
BC171682.1 - 1 - 2 92 67
BC171686.1 - 1 - 2 92 67

View Full Product Details