Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c1qbp
Entrez Gene ID
334619
Gene Name
complement component 1, q subcomponent binding protein
Gene Aliases
fa14h03, sb:cb785, si:ch1073-329n19.2, wu:fa14h03, zgc:110137
Chromosome Location
Chr.5: 3914402 - 3927763 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017858.2 NP_001017858.2 3 - 4 566 84
GenBank mRNA BC092751.1 - 3 - 4 513 84
BC164455.1 - 3 - 4 480 84
EH440105.1 - 3 - 4 539 84

View Full Product Details