Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myl10
Entrez Gene ID
550569
Gene Name
myosin, light chain 10, regulatory
Gene Aliases
id:ibd2025, mlc10, zgc:110679
Chromosome Location
Chr.10: 33171862 - 33179915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017871.1 NP_001017871.1 5 - 6 402 82
GenBank mRNA BC092977.1 - 5 - 6 402 82

View Full Product Details