The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017902.1 | NP_001017902.1 | 2 - 3 | 252 | 115 |
GenBank mRNA | BC092943.1 | - | 2 - 3 | 252 | 115 |
BC154164.1 | - | 2 - 3 | 256 | 115 |