Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mipb
Entrez Gene ID
553420
Gene Name
major intrinsic protein of lens fiber b
Gene Aliases
mip2, zgc:110015
Chromosome Location
Chr.23: 6637611 - 6641218 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020520.1 NP_001018356.1 1 - 2 812 78
GenBank mRNA BC098535.1 - 1 - 2 823 78
DQ003080.1 - 1 - 2 812 78
FJ655389.1 - 1 - 2 395 78

View Full Product Details