Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eri1
Entrez Gene ID
553641
Gene Name
exoribonuclease 1
Gene Aliases
thex1, zgc:110635
Chromosome Location
Chr.21: 19907763 - 19915902 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020614.1 NP_001018450.1 5 - 6 927 129
GenBank mRNA BC095343.1 - 5 - 6 927 129
BC153552.1 - 5 - 6 797 129
BC164140.1 - 5 - 6 692 129

View Full Product Details