Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stxbp6l
Entrez Gene ID
100002110
Gene Name
syntaxin binding protein 6 (amisyn), like
Gene Aliases
si:dkey-29m11.7, si:dkey-63j12.1, zgc:110123
Chromosome Location
Chr.20: 200714 - 214739 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020738.2 NP_001018574.2 2 - 3 287 132
XM_005160403.4 XP_005160460.1 4 - 5 772 132
XM_009294576.3 XP_009292851.1 4 - 5 773 132
XM_021468334.1 XP_021324009.1 3 - 4 641 132
GenBank mRNA BC095177.1 - 2 - 3 287 132

View Full Product Details