Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ankrd29
Entrez Gene ID
553798
Gene Name
ankyrin repeat domain 29
Gene Aliases
si:dkey-121a11.5, zgc:111996
Chromosome Location
Chr.22: 16418695 - 16431136 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020760.1 NP_001018596.1 8 - 9 978 73
XM_005161708.4 XP_005161765.1 8 - 9 860 73
GenBank mRNA BC095639.1 - 8 - 9 978 73

View Full Product Details