Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fdps
Entrez Gene ID
552997
Gene Name
farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase)
Gene Aliases
fb23g06, im:6904202, wu:fb23g06
Chromosome Location
Chr.16: 14010173 - 14022911 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025471.1 NP_001020642.1 7 - 8 736 141
GenBank mRNA BC083515.1 - 6 - 7 667 141
BC097112.1 - 7 - 8 736 141
BC164398.1 - 6 - 7 692 141

View Full Product Details