Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs7bpb
Entrez Gene ID
337556
Gene Name
regulator of G protein signaling 7 binding protein b
Gene Aliases
fj83a05, rgs7bp, wu:fj83a05, zgc:110002
Chromosome Location
Chr.21: 20933383 - 20939295 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030069.1 NP_001025240.1 5 - 6 865 73
GenBank mRNA BC096790.1 - 5 - 6 865 73
BC152595.1 - 5 - 6 843 73
BC153404.1 - 5 - 6 569 73
BC165928.1 - 5 - 6 696 73

View Full Product Details