The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098195.1 | NP_001091665.1 | 3 - 4 | 386 | 61 |
GenBank mRNA | BC139661.1 | - | 3 - 4 | 386 | 61 |