Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrps22
Entrez Gene ID
556778
Gene Name
mitochondrial ribosomal protein S22
Gene Aliases
zgc:165596
Chromosome Location
Chr.15: 34181 - 58364 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098735.1 NP_001092205.1 2 - 3 293 72
GenBank mRNA BC142888.1 - 2 - 3 293 72
BC150427.1 - 2 - 3 437 72

View Full Product Details