Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agbl2
Entrez Gene ID
568152
Gene Name
ATP/GTP binding protein-like 2
Gene Aliases
zgc:165648, zte25
Chromosome Location
Chr.25: 28596989 - 28638145 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320148.1 NP_001307077.1 3 - 4 463 132
XM_017354091.2 XP_017209580.1 3 - 4 506 132
GenBank mRNA BC146747.1 - 3 - 4 459 132

View Full Product Details