Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hcst
Entrez Gene ID
100101460
Gene Name
hematopoietic cell signal transducer
Gene Aliases
dap10
Chromosome Location
Chr.16: 25184209 - 25188121 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100102.2 NP_001093572.2 3 - 4 153 69
GenBank mRNA EF158445.1 - 3 - 4 140 69
EH454362.1 - 3 - 4 139 69

View Full Product Details