Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
capns1b
Entrez Gene ID
560033
Gene Name
calpain, small subunit 1 b
Gene Aliases
cb536, fa18a08, wu:fa18a08, zgc:171735
Chromosome Location
Chr.15: 29116114 - 29122830 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109706.1 NP_001103176.1 9 - 10 736 134
XM_005157735.1 XP_005157792.1 9 - 10 676 134
GenBank mRNA BC151952.1 - 9 - 10 736 134
BC162123.1 - 9 - 10 722 134
BC162479.1 - 9 - 10 722 134

View Full Product Details