Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2aa4
Entrez Gene ID
415182
Gene Name
cytochrome P450, family 2, subfamily AA, polypeptide 4
Gene Aliases
cyp2, zgc:86915
Chromosome Location
Chr.23: 42362337 - 42378392 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002092.2 NP_001002092.1 2 - 3 374 71
XM_017353539.2 XP_017209028.1 2 - 3 367 71
GenBank mRNA AB303423.1 - 2 - 3 371 71
BC071533.1 - 2 - 3 376 71

View Full Product Details