The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002113.1 | NP_001002113.1 | 6 - 7 | 698 | 69 |
GenBank mRNA | BC071481.1 | - | 6 - 7 | 698 | 69 |