Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cav2
Entrez Gene ID
415240
Gene Name
caveolin 2
Gene Aliases
wu:fc52e01, zgc:86696
Chromosome Location
Chr.25: 18556606 - 18561795 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002150.1 NP_001002150.1 2 - 3 403 63
GenBank mRNA BC071376.1 - 2 - 3 403 63
BC154297.1 - 2 - 3 421 63
DQ015874.1 - 2 - 3 366 63

View Full Product Details