Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhrs1
Entrez Gene ID
368670
Gene Name
dehydrogenase/reductase (SDR family) member 1
Gene Aliases
si:zc14a17.8, wu:fa91c02, zgc:91825
Chromosome Location
Chr.2: 305336 - 313447 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002205.1 NP_001002205.1 2 - 3 192 78
GenBank mRNA BC072703.1 - 2 - 3 192 78
BC164751.1 - 1 - 2 182 78

View Full Product Details