Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abca4a
Entrez Gene ID
798993
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 4a
Gene Aliases
zgc:91823
Chromosome Location
Chr.24: 28561038 - 28575398 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002206.1 NP_001002206.1 5 - 6 581 94
GenBank mRNA BC074030.1 - 5 - 6 581 94
BC164633.1 - 4 - 5 477 94

View Full Product Details