Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rcn2
Entrez Gene ID
571340
Gene Name
reticulocalbin 2
Gene Aliases
si:dkey-24l11.6, zgc:110594
Chromosome Location
Chr.18: 151778 - 163128 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030263.1 NP_001025434.1 3 - 4 303 108
GenBank mRNA BC098559.1 - 3 - 4 303 108

View Full Product Details