Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cst3
Entrez Gene ID
791878
Gene Name
cystatin C (amyloid angiopathy and cerebral hemorrhage)
Gene Aliases
fb51d07, wu:fb24g06, wu:fb51d07, wu:fc55f03, zgc:136227
Chromosome Location
Chr.13: 33681873 - 33683913 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031673.2 NP_001026843.2 2 - 3 415 112
GenBank mRNA BC117569.1 - 2 - 3 374 112
BC165901.1 - 2 - 3 339 112
DQ118400.1 - 2 - 3 363 112
EB868200.1 - 2 - 3 417 112

View Full Product Details