Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
insl5a
Entrez Gene ID
566602
Gene Name
insulin-like 5a
Gene Aliases
RFLA1, insl5, rlx3b
Chromosome Location
Chr.6: 177707 - 181573 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037669.1 NP_001032758.1 1 - 2 184 147
GenBank mRNA AY834224.1 - 1 - 2 184 147
BC163160.1 - 1 - 2 147
BC163186.1 - 1 - 2 147
EU437466.1 - 1 - 2 147

View Full Product Details