Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inha
Entrez Gene ID
570520
Gene Name
inhibin subunit alpha
Gene Aliases
si:dkey-91f15.2
Chromosome Location
Chr.6: 296469 - 320610 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045204.1 NP_001038669.1 1 - 2 307 62

View Full Product Details