Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- inppl1b
- Entrez Gene ID
-
368433
- Gene Name
- inositol polyphosphate phosphatase-like 1b
- Gene Aliases
-
inppl1, ship-2b, si:bz46j2.7, si:rp71-46j2.7
- Chromosome Location
-
Chr.14: 32793586 - 32824407 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DQ272662.1 |
- |
8 - 9 |
947 |
65 |
View Full Product Details