Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btr02
Entrez Gene ID
562439
Gene Name
bloodthirsty-related gene family, member 2
Gene Aliases
wu:fc66g05, zgc:153136
Chromosome Location
Chr.3: 50128266 - 50139846 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077158.2 NP_001070626.2 3 - 4 446 146
GenBank mRNA BC122154.1 - 3 - 4 446 146

View Full Product Details