Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
agmat
Entrez Gene ID
556522
Gene Name
agmatine ureohydrolase (agmatinase)
Gene Aliases
auh, zgc:153353
Chromosome Location
Chr.23: 37482727 - 37490252 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077551.1 NP_001071019.1 2 - 3 646 82
GenBank mRNA BC124326.1 - 2 - 3 646 82
BC164767.1 - 2 - 3 549 82

View Full Product Details