Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
agmat
Entrez Gene ID
556522
Gene Name
agmatine ureohydrolase (agmatinase)
Gene Aliases
auh, zgc:153353
Chromosome Location
Chr.23: 37482727 - 37490252 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077551.1 NP_001071019.1 5 - 6 1080 96
GenBank mRNA BC124326.1 - 5 - 6 1080 96
BC164767.1 - 5 - 6 983 96

View Full Product Details