The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077584.1 | NP_001071052.1 | 5 - 6 | 673 | 71 |
GenBank mRNA | BC124172.1 | - | 5 - 6 | 673 | 71 |
BI326762.1 | - | 2 - 3 | 138 | 71 |