Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spoplb
Entrez Gene ID
558170
Gene Name
speckle type BTB/POZ protein like b
Gene Aliases
im:6961637, zgc:153365
Chromosome Location
Chr.11: 33818302 - 33842711 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079969.1 NP_001073438.1 8 - 9 1019 109
GenBank mRNA BC125865.1 - 8 - 9 1019 109

View Full Product Details