Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
palm3
Entrez Gene ID
790940
Gene Name
paralemmin 3
Gene Aliases
zgc:158252
Chromosome Location
Chr.6: 7977495 - 8075517 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080085.1 NP_001073554.1 3 - 4 516 93
XM_021476937.1 XP_021332612.1 3 - 4 633 93
GenBank mRNA BC128825.1 - 3 - 4 516 93

View Full Product Details