Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp3ccb
Entrez Gene ID
565241
Gene Name
protein phosphatase 3, catalytic subunit, gamma isozyme, b
Gene Aliases
im:7163784, zgc:158685
Chromosome Location
Chr.10: 222970 - 264483 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080594.2 NP_001074063.2 10 - 11 1459 108
GenBank mRNA BC129382.1 - 10 - 11 1450 108
BC165627.1 - 10 - 11 1180 108

View Full Product Details