Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppp3ccb
Entrez Gene ID
565241
Gene Name
protein phosphatase 3, catalytic subunit, gamma isozyme, b
Gene Aliases
im:7163784, zgc:158685
Chromosome Location
Chr.10: 222970 - 264483 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080594.2 NP_001074063.2 6 - 7 1083 103
XM_005155313.4 XP_005155370.1 6 - 7 1114 103
XM_005155314.4 XP_005155371.1 6 - 7 1114 103
GenBank mRNA BC129382.1 - 6 - 7 1074 103
BC165627.1 - 6 - 7 804 103

View Full Product Details