Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
suv39h1b
Entrez Gene ID
326906
Gene Name
suppressor of variegation 3-9 homolog 1b
Gene Aliases
fe47h06, wu:fb18a10, wu:fe47h06, zgc:136593
Chromosome Location
Chr.8: 25750963 - 25761544 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126482.1 NP_001119954.1 3 - 4 1269 64
GenBank mRNA BC052225.1 - 3 - 4 881 64
BC115190.1 - 3 - 4 876 64

View Full Product Details