Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp4r2b
Entrez Gene ID
100005419
Gene Name
protein phosphatase 4, regulatory subunit 2b
Gene Aliases
wu:fa17h08, wu:fc23g05
Chromosome Location
Chr.6: 80576 - 90898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110371.1 NP_001103841.1 3 - 4 366 98
GenBank mRNA BC129134.1 - 3 - 4 350 98
BC154497.1 - 3 - 4 366 98

View Full Product Details