Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mri1
Entrez Gene ID
100002302
Gene Name
methylthioribose-1-phosphate isomerase 1
Gene Aliases
CH73-131E21.1, M1Pi, zgc:172216
Chromosome Location
Chr.1: 55191150 - 55210619 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111238.1 NP_001104708.1 1 - 2 236 80
GenBank mRNA BC155626.1 - 1 - 2 236 80
BC171374.1 - 1 - 2 200 80
BC171376.1 - 1 - 2 80

View Full Product Details