Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myh7
Entrez Gene ID
30616
Gene Name
myosin heavy chain 7
Gene Aliases
mg:cb02g011, smyhc3, vmhc
Chromosome Location
Chr.2: 24256521 - 24269913 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112733.1 NP_001106204.1 24 - 25 3321 86
GenBank mRNA AF114427.2 - 24 - 25 3327 86

View Full Product Details