Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spred3
Entrez Gene ID
568925
Gene Name
sprouty related EVH1 domain containing 3
Gene Aliases
si:ch211-255l12.2
Chromosome Location
Chr.18: 44596787 - 44611301 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113215.2 NP_001106686.2 2 - 3 203 116
XM_005159247.4 XP_005159304.1 2 - 3 501 116
XM_021467687.1 XP_021323362.1 2 - 3 292 116
XM_021467689.1 XP_021323364.1 2 - 3 292 116
GenBank mRNA BC155342.1 - 2 - 3 203 116

View Full Product Details