Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
keap1b
Entrez Gene ID
100003679
Gene Name
kelch-like ECH-associated protein 1b
Gene Aliases
zgc:171654
Chromosome Location
Chr.6: 131998 - 138620 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113477.1 NP_001106948.1 4 - 5 1499 81
XM_005168685.3 XP_005168742.1 4 - 5 1647 81
XM_021476739.1 XP_021332414.1 4 - 5 1537 81
GenBank mRNA AB271119.1 - 4 - 5 1499 81

View Full Product Details