Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccl20b
Entrez Gene ID
563152
Gene Name
chemokine (C-C motif) ligand 20b
Gene Aliases
ccl20, si:dkey-150o13.1
Chromosome Location
Chr.24: 26006918 - 26009130 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113595.1 NP_001107067.1 1 - 2 69 79
GenBank mRNA AB331755.1 - 1 - 2 69 79

View Full Product Details